missing translation for 'onlineSavingsMsg'
Learn More

(2-Butenyl)triphenylphosphonium chloride, 97%

Catalog No. AAL0050206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0050206 5 g
AAL0050214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0050206 Supplier Thermo Scientific Chemicals Supplier No. L0050206

CAS: 13138-25-5 | C22H22ClP | 352.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13138-25-5
Molecular Formula C22H22ClP
Molecular Weight (g/mol) 352.84
MDL Number MFCD00051868
InChI Key YYTDJYJBYMQMDI-SQQVDAMQSA-M
Synonym 2-butenyl triphenylphosphonium chloride, but-2-en-1-yltriphenylphosphonium chloride, 2e-but-2-en-1-yltriphenylphosphanium chloride
PubChem CID 11824356
IUPAC Name [(E)-but-2-enyl]-triphenylphosphanium;chloride
SMILES [Cl-].C\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽225°C to 230°C (decomposition)
Quantity 5 g
Beilstein 4170527
Sensitivity Hygroscopic
Formula Weight 352.84
Percent Purity 97%
Chemical Name or Material (2-Butenyl)triphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only