missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotriphenylene 98.0+%, TCI America™

Catalog No. B44421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44421G 1 g
B44425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44421G Supplier TCI America Supplier No. B44421G
Only null left

Chemical Identifiers

CAS 19111-87-6
Molecular Formula C18H11Br
Molecular Weight (g/mol) 307.19
MDL Number MFCD16659043
InChI Key GEDOYYDMCZUHNW-UHFFFAOYSA-N
PubChem CID 18381174
IUPAC Name 2-bromotriphenylene
SMILES BrC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 307.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromotriphenylene