missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromotoluene 98.0+%, TCI America™
$42.36 - $365.79
Chemical Identifiers
| CAS | 95-46-5 |
|---|---|
| Molecular Formula | C7H7Br |
| Molecular Weight (g/mol) | 171.04 |
| MDL Number | MFCD00000068 |
| InChI Key | QSSXJPIWXQTSIX-UHFFFAOYSA-N |
| Synonym | 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide |
| PubChem CID | 7236 |
| IUPAC Name | 1-bromo-2-methylbenzene |
| SMILES | CC1=CC=CC=C1Br |
Chemical Identifiers
| 95-46-5 | |
| 171.04 | |
| QSSXJPIWXQTSIX-UHFFFAOYSA-N | |
| 7236 | |
| CC1=CC=CC=C1Br |
| C7H7Br | |
| MFCD00000068 | |
| 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide | |
| 1-bromo-2-methylbenzene |
Specifications
| 95-46-5 | |
| Yellow | |
| C7H7Br | |
| 25 g | |
| QSSXJPIWXQTSIX-UHFFFAOYSA-N | |
| 1-bromo-2-methylbenzene | |
| 7236 | |
| ≥98.0% (GC) | |
| 2-Bromotoluene |
| -26°C | |
| 181°C | |
| MFCD00000068 | |
| 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide | |
| CC1=CC=CC=C1Br | |
| 171.04 | |
| 171.04 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0088
RTECSNumber : XS7965500
TSCA : Yes