missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotoluene 98.0+%, TCI America™

Catalog No. B0659500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0659500G 500 g
B065925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0659500G Supplier TCI America Supplier No. B0659500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-46-5
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000068
InChI Key QSSXJPIWXQTSIX-UHFFFAOYSA-N
Synonym 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
PubChem CID 7236
IUPAC Name 1-bromo-2-methylbenzene
SMILES CC1=CC=CC=C1Br

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 181°C
Quantity 500 g
Formula Weight 171.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromotoluene
EINECSNumber (3)-0088
RTECSNumber XS7965500
TSCA Yes