missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophenol, 97%

Catalog No. AC221980010
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221980010 1 g Glass bottle
AC221980100 10 g Glass bottle
AC221980500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221980010 Supplier Thermo Scientific Chemicals Supplier No. 221980010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6320-02-1 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6320-02-1
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004827
InChI Key YUQUNWNSQDULTI-UHFFFAOYSA-N
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
PubChem CID 80599
SMILES SC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6040g/mL
Boiling Point 114°C to 116°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6345 to 1.6365
Linear Formula BrC6H4SH
Quantity 1 g
Beilstein 06, II, 300
Specific Gravity 1.604
Solubility Information Solubility in water: immiscible
IUPAC Name 2-chlorobenzenethiol
Formula Weight 189.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 228-665-4

RUO – Research Use Only