missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophenol, 97%

Catalog No. p-7028300
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2553706 5 g
AAB2553714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2553706 Supplier Thermo Scientific Chemicals Supplier No. B2553706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6320-02-1 | C6H5ClS | 144.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6320-02-1
MDL Number MFCD00004827

Specifications

Density 1.604
Boiling Point 114°C to 116°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Molecular Formula C6H5ClS
Refractive Index 1.635
Quantity 5 g
UN Number UN2922
Beilstein 2039769
Sensitivity Air Sensitive
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)S)Cl
IUPAC Name 2-chlorobenzenethiol
Molecular Weight (g/mol) 144.62
PubChem CID 80599
Formula Weight 189.08
Percent Purity 97%
Chemical Name or Material 2-Bromothiophenol
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 228-665-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only