missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole 96.0+%, TCI America™

Catalog No. p-7119429
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20235G 5 g
B202325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20235G Supplier TCI America Supplier No. B20235G
Only null left

Chemical Identifiers

CAS 19614-16-5
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00000066
InChI Key ALAQDUSTXPEHMH-UHFFFAOYSA-N
Synonym 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
PubChem CID 88166
IUPAC Name 1-bromo-2-methylsulfanylbenzene
SMILES CSC1=CC=CC=C1Br

Specifications

Color Brown
Boiling Point 256°C
Quantity 5 g
Formula Weight 203.10
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromothioanisole