missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole, 98%

CAS: 19614-16-5 | C7H7BrS | 203.097 g/mol

Supplier:  Thermo Scientific Chemicals A1363206

 View more versions of this product

Catalog No. AAA1363206

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromothioanisole
-24°C
140°C to 141°C (20 mmHg)
C7H7BrS
MFCD00000066
2243716
ALAQDUSTXPEHMH-UHFFFAOYSA-N
1-bromo-2-methylsulfanylbenzene
88166
98%
19614-16-5
1.524
>110°C (230°F)
1.633
5 g
2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
CSC1=CC=CC=C1Br
203.097
203.11

Chemical Identifiers

19614-16-5
203.097
ALAQDUSTXPEHMH-UHFFFAOYSA-N
88166
CSC1=CC=CC=C1Br
C7H7BrS
MFCD00000066
2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
1-bromo-2-methylsulfanylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 243-183-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only