missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole, 98%

Catalog No. AAA1363203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363203 1 g
AAA1363206 5 g
AAA1363214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1363203 Supplier Thermo Scientific Chemicals Supplier No. A1363203

CAS: 19614-16-5 | C7H7BrS | 203.097 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19614-16-5
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00000066
InChI Key ALAQDUSTXPEHMH-UHFFFAOYSA-N
Synonym 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
PubChem CID 88166
IUPAC Name 1-bromo-2-methylsulfanylbenzene
SMILES CSC1=CC=CC=C1Br

Specifications

Melting Point -24°C
Density 1.524
Boiling Point 140°C to 141°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.633
Quantity 1 g
Beilstein 2243716
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 2-Bromothioanisole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 243-183-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only