missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. B45781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45781G 1 g
B45785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45781G Supplier TCI America Supplier No. B45781G

Chemical Identifiers

CAS 5198-88-9
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.029
MDL Number MFCD04115729
InChI Key BEGREHRAUWCAHV-UHFFFAOYSA-N
Synonym 2-bromo-4-thiazolecarboxylic acid, 2-bromothiazole-4-carboxylic acid, 2-bromo-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-bromo, 2-bromo-4-thiazolecarboxylicacid, 2-bromothiazole-4-carboxylicacid, 2-bromo-4-carboxy-1,3-thiazole, pubchem22900, acmc-1avkr, 2-bromo-4-carboxythiazole
PubChem CID 2763209
IUPAC Name 2-bromo-1,3-thiazole-4-carboxylic acid
SMILES C1=C(N=C(S1)Br)C(=O)O

Specifications

Melting Point 217°C
Color White
Quantity 1 g
Formula Weight 208.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromothiazole-4-carboxylic Acid