accessibility menu, dialog, popup

UserName

OOSBrN

CAS RN 5198-88-9

IUPAC Name: 2-bromo-1,3-thiazole-4-carboxylate
Synonyms: 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 207.02
Molecular Formula: C4HBrNO2S
InChi Key: BEGREHRAUWCAHV-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(Br)=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)