missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotetradecane, 95%

Catalog No. AAL1097406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1097406 5 g
AAL1097414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1097406 Supplier Thermo Scientific Chemicals Supplier No. L1097406

CAS: 74036-95-6 | C14H29Br | 277.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74036-95-6
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00045033
InChI Key WZGQIDWFBFDMLE-UHFFFAOYSA-N
Synonym tetradecane, 2-bromo
PubChem CID 12798926
IUPAC Name 2-bromotetradecane
SMILES CCCCCCCCCCCCC(C)Br

Specifications

Boiling Point 160°C to 161°C (8 mmHg)
Refractive Index 1.46
Quantity 5 g
Beilstein 2071530
Formula Weight 277.3
Percent Purity 95%
Chemical Name or Material 2-Bromotetradecane
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only