Learn More
2-Bromoterephthalic acid, 97%
CAS: 586-35-6 | C8H5BrO4 | 245.028 g/mol
Supplier: Thermo Scientific Chemicals A1439322
Description
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| 2-Bromoterephthalic acid | |
| 297°C to 300°C | |
| MFCD00002403 | |
| UN2811 | |
| bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9 | |
| QPBGNSFASPVGTP-UHFFFAOYSA-N | |
| 2-bromoterephthalic acid | |
| 68513 | |
| 97% |
| 586-35-6 | |
| C8H5BrO4 | |
| 100 g | |
| 2616163 | |
| Soluble in water. | |
| C1=CC(=C(C=C1C(=O)O)Br)C(=O)O | |
| 245.028 | |
| 245.03 |
Chemical Identifiers
| 586-35-6 | |
| 245.028 | |
| QPBGNSFASPVGTP-UHFFFAOYSA-N | |
| 68513 | |
| C1=CC(=C(C=C1C(=O)O)Br)C(=O)O |
| C8H5BrO4 | |
| MFCD00002403 | |
| bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9 | |
| 2-bromoterephthalic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 209-572-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only