missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoquinoxaline 98.0+%, TCI America™

Numéro de catalogue. B4632200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B4632200MG 200 mg
B46321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B4632200MG Fournisseur TCI America Code fournisseur B4632200MG
Il en reste null

Identifiants chimiques

CAS 36856-91-4
Molecular Formula C8H5BrN2
Molecular Weight (g/mol) 209.046
MDL Number MFCD07368206
InChI Key XVKLMYQMVPHUPN-UHFFFAOYSA-N
Synonym quinoxaline, 2-bromo, bromoquinoxaline, 2-bromoquinoxaline, ksc226a0j
PubChem CID 582225
IUPAC Name 2-bromoquinoxaline
SMILES C1=CC=C2C(=C1)N=CC(=N2)Br

Spécifications

Melting Point 57°C
Quantity 200 mg
Formula Weight 209.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoquinoxaline