accessibility menu, dialog, popup

UserName

CAS RN 36856-91-4

NNBr

CAS RN 36856-91-4

IUPAC Name: 2-bromoquinoxaline
Synonyms: 2-bromoquinoxaline
Molecular Weight (g/mol): 209.05
Molecular Formula: C8H5BrN2
InChi Key: XVKLMYQMVPHUPN-UHFFFAOYSA-N
SMILES: BrC1=NC2=CC=CC=C2N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results