missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-4-carboxylic acid, 97%

Catalog No. p-7047859
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1985503 1 g
AAL1985506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1985503 Supplier Thermo Scientific Chemicals Supplier No. L1985503
Only null left

CAS: 66572-56-3 | C6H4BrNO2 | 202.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66572-56-3
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646069
InChI Key YBTKGKVQEXAYEM-UHFFFAOYSA-N
Synonym 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt
PubChem CID 222701
IUPAC Name 2-bromopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)Br

Specifications

Melting Point ∽225°C (decomposition)
Quantity 1 g
Beilstein 471895
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only