missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-4-carboxamide, 97%

Catalog No. p-7047858
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1985403 1 g
AAL1985406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1985403 Supplier Thermo Scientific Chemicals Supplier No. L1985403
Only null left

CAS: 29840-73-1 | C6H5BrN2O | 201.023 g/mol

2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 29840-73-1
Molecular Formula C6H5BrN2O
Molecular Weight (g/mol) 201.023
MDL Number MFCD00234156
InChI Key KCELTDZFDHPTBI-UHFFFAOYSA-N
PubChem CID 4625407
IUPAC Name 2-bromopyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Br

Specifications

Melting Point 192°C
Quantity 1 g
Beilstein 471892
Synonym 2-bromoisonicotinamide, 2-bromo-isonicotinamide, 2-bromo-4-pyridinecarboxamide, 2-bromopyridin-4-ylcarboxamide, 4-pyridinecarboxamide, 2-bromo, 2-bromo-isonicotimide, 4-pyridinecarboxamide,2-bromo, pubchem16641, acmc-1cr70, 2-bromanylpyridine-4-carboxamide
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-4-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only