CAS RN 29840-73-1
CAS RN 29840-73-1
IUPAC Name:
2-bromopyridine-4-carboxamide
Synonyms:
2-bromopyridine-4-carboxamide
Molecular Weight (g/mol):
201.02
Molecular Formula:
C6H5BrN2O
InChi Key:
KCELTDZFDHPTBI-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=NC(Br)=C1
1
–
1
of
1
results
1
2-Bromopyridine-4-carboxamide, 97%
CAS: 29840-73-1 Molecular Formula: C6H5BrN2O Molecular Weight (g/mol): 201.023 MDL Number: MFCD00234156 InChI Key: KCELTDZFDHPTBI-UHFFFAOYSA-N PubChem CID: 4625407 IUPAC Name: 2-bromopyridine-4-carboxamide SMILES: C1=CN=C(C=C1C(=O)N)Br