missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropane 99.0+%, TCI America™

Catalog No. p-7118582
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B063925G 25 g
B0639500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B063925G Supplier TCI America Supplier No. B063925G

Chemical Identifiers

CAS 75-26-3
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000147
InChI Key NAMYKGVDVNBCFQ-UHFFFAOYSA-N
Synonym isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan
PubChem CID 6358
IUPAC Name 2-bromopropane
SMILES CC(C)Br

Specifications

Melting Point -89°C
Color Colorless
Boiling Point 59°C
Quantity 25 g
UN Number 2344
Formula Weight 122.99
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromopropane
EINECSNumber (2)-0076
RTECSNumber TX4111000
TSCA Yes