missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropane, 99%

Catalog No. p-4229114
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107322500 250 mL Glass bottle
AC107320010 1 L Glass bottle
AC107320025 2.5 L Glass bottle
AC107320100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC107322500 Supplier Thermo Scientific Chemicals Supplier No. 107322500
Only null left

CAS: 75-26-3 | C3H7Br | 122.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-26-3
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.99
InChI Key NAMYKGVDVNBCFQ-UHFFFAOYSA-N
Synonym isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan
PubChem CID 6358
IUPAC Name 2-bromopropane
SMILES CC(C)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -89.0°C
Color Brown to Colorless
Density 1.3100g/mL
Boiling Point 59.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270
Linear Formula (CH3)2CHBr
Quantity 250 mL
Beilstein 01,108
Fieser 09,69
Merck Index 15,5256
Specific Gravity 1.31
Solubility Information Solubility in water: 0.3g/100mL. Other solubilities: miscible with alcohol,benzene,chloroform,ether,and acetone
Formula Weight 122.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
May damage fertility.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-855-1

RUO – Research Use Only