missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropane, 99%

Catalog No. AAA12944AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12944AU 1000 mL
AAA12944AK 250 mL
AAA129440F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12944AU Supplier Thermo Scientific Chemicals Supplier No. A12944AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75-26-3 | C3H7Br | 122.993 g/mol

2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water and acetone.

Notes
Avoid all possible sources of ignition.Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 75-26-3
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000147
InChI Key NAMYKGVDVNBCFQ-UHFFFAOYSA-N
PubChem CID 6358
IUPAC Name 2-bromopropane
SMILES CC(C)Br

Specifications

Melting Point -89°C
Density 1.31
Boiling Point 59°C to 60°C
Flash Point 19°C (66°F)
Assay Percent Range 99%
Refractive Index 1.4255
Linear Formula CH3CHBrCH3
Quantity 1000 mL
UN Number UN2344
Beilstein 741852
Merck Index 14,5210
Synonym isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan
Solubility Information Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water and acetone.
Formula Weight 123
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopropane
Show More Show Less
Hazard Category H225-H335-H336-H360F-H373
Hazard Statement GHS H Statement
H225-H360-H373
Highly flammable liquid and vapour.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES
EINECSNumber 200-855-1
RTECSNumber TX4111000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only