missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylhydrazine hydrochloride, 94%

Catalog No. p-7027307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2396606 5 g
AAB2396614 25 g
AAB2396622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2396606 Supplier Thermo Scientific Chemicals Supplier No. B2396606
Only null left

CAS: 50709-33-6 | C6H8BrClN2 | 223.498 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50709-33-6
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.498
MDL Number MFCD00012926
InChI Key PHCYUJRYSFMJMG-UHFFFAOYSA-N
Synonym 2-bromophenylhydrazine hydrochloride, 2-bromophenyl hydrazine hydrochloride, 2-bromophenylhydrazine hcl, hydrazine, 2-bromophenyl-, monohydrochloride, 1-2-bromophenyl hydrazine hydrochloride, 1-bromo-2-hydrazino benzene hydrochloride, pubchem4463, acmc-1b1zz, ksc270g5h, 2-bromophenylhydrazine, chloride
PubChem CID 2723912
IUPAC Name (2-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)NN)Br.Cl

Specifications

Melting Point ∼188°C (decomposition)
Quantity 5 g
Formula Weight 223.51
Percent Purity 95%
Chemical Name or Material 2-Bromophenylhydrazine hydrochloride
Hazard Category H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 256-728-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only