missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromophenylethynyl)trimethylsilane 98.0+%, TCI America™

Catalog No. p-7123979
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B51381G 1 g
B51385G 5 g
2 options

Catalog No. B51381G

Supplier: TCI America B51381G

Only null left

Chemical Identifiers

CAS 38274-16-7
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01321391
InChI Key FABNGXSCLHXUOH-UHFFFAOYSA-N
Synonym 2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
PubChem CID 4547398
IUPAC Name [2-(2-bromophenyl)ethynyl]trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=CC=C1Br

Specifications

Boiling Point 91°C
Quantity 1 g
Formula Weight 253.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromophenylethynyl)trimethylsilane
TSCA No
Recommended Storage Refrigerator