missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromophenylethynyl)trimethylsilane 98.0+%, TCI America™

Catalog No. B51385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51385G 5 g
B51381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51385G Supplier TCI America Supplier No. B51385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38274-16-7
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01321391
InChI Key FABNGXSCLHXUOH-UHFFFAOYSA-N
Synonym 2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
PubChem CID 4547398
IUPAC Name [2-(2-bromophenyl)ethynyl]trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=CC=C1Br

Specifications

Boiling Point 91°C
Quantity 5 g
Formula Weight 253.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromophenylethynyl)trimethylsilane
TSCA No
Recommended Storage Refrigerator