missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetone 98.0+%, TCI America™

Catalog No. B38425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38425G 5 g
B38421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38425G Supplier TCI America Supplier No. B38425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21906-31-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
InChI Key TZIAZLUAMDLDJF-UHFFFAOYSA-N
Synonym 2-bromophenylacetone, 1-2-bromophenyl propan-2-one, 2-bromophenyl acetone, 1-2-bromophenyl-2-propanone, 1-acetonyl-2-bromobenzene, 2-propanone, 1-2-bromophenyl, o-bromo benyl methyl ketone, 1-2-bromophenyl acetone, o-bromo benzyl methyl ketone, pubchem23388
PubChem CID 2734092
IUPAC Name 1-(2-bromophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromophenylacetone