missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetic Acid 98.0+%, TCI America™

Catalog No. B19575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19575G 5 g
B195725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19575G Supplier TCI America Supplier No. B19575G

Chemical Identifiers

CAS 18698-97-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00004314
InChI Key DWXSYDKEWORWBT-UHFFFAOYSA-N
Synonym 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
PubChem CID 87754
IUPAC Name 2-(2-bromophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Br

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromophenylacetic Acid