Learn More
2-Bromophenylacetic acid, 98+%
CAS: 18698-97-0 | C8H7BrO2 | 215.046 g/mol
Supplier: Thermo Scientific Chemicals A1690014
Description
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a pharmaceutical intermediate. It is also used in organic synthesis.
Solubility
Very faint turbidity in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
| 2-Bromophenylacetic acid | |
| 105°C to 106°C | |
| MFCD00004314 | |
| 2250655 | |
| Very faint turbidity in methanol. | |
| C1=CC=C(C(=C1)CC(=O)O)Br | |
| 215.046 | |
| 215.05 |
| 18698-97-0 | |
| C8H7BrO2 | |
| 25 g | |
| 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg | |
| DWXSYDKEWORWBT-UHFFFAOYSA-N | |
| 2-(2-bromophenyl)acetic acid | |
| 87754 | |
| ≥98% |
Chemical Identifiers
| 18698-97-0 | |
| 215.046 | |
| DWXSYDKEWORWBT-UHFFFAOYSA-N | |
| 87754 | |
| C1=CC=C(C(=C1)CC(=O)O)Br |
| C8H7BrO2 | |
| MFCD00004314 | |
| 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg | |
| 2-(2-bromophenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 242-509-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only