missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetic acid, 98+%

Catalog No. AAA1690006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1690006 5 g
AAA1690014 25 g
AAA1690022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1690006 Supplier Thermo Scientific Chemicals Supplier No. A1690006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18698-97-0 | C8H7BrO2 | 215.046 g/mol

Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

Solubility
Very faint turbidity in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 18698-97-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00004314
InChI Key DWXSYDKEWORWBT-UHFFFAOYSA-N
Synonym 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
PubChem CID 87754
IUPAC Name 2-(2-bromophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Br

Specifications

Melting Point 105°C to 106°C
Quantity 5 g
Beilstein 2250655
Solubility Information Very faint turbidity in methanol.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 2-Bromophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-509-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only