missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenyl Methyl Sulfoxide 98.0+%, TCI America™
$194.44 - $581.41
Chemical Identifiers
| CAS | 7321-58-6 |
|---|---|
| Molecular Formula | C7H7BrOS |
| Molecular Weight (g/mol) | 219.096 |
| MDL Number | MFCD18089235 |
| InChI Key | NNNHMHLNXXGWBA-UHFFFAOYSA-N |
| Synonym | 1-Bromo-2-(methylsulfinyl)benzene |
| PubChem CID | 11074873 |
| IUPAC Name | 1-bromo-2-methylsulfinylbenzene |
| SMILES | CS(=O)C1=CC=CC=C1Br |
Chemical Identifiers
| 7321-58-6 | |
| 219.096 | |
| NNNHMHLNXXGWBA-UHFFFAOYSA-N | |
| 11074873 | |
| CS(=O)C1=CC=CC=C1Br |
| C7H7BrOS | |
| MFCD18089235 | |
| 1-Bromo-2-(methylsulfinyl)benzene | |
| 1-bromo-2-methylsulfinylbenzene |
Specifications
| 7321-58-6 | |
| 109°C | |
| MFCD18089235 | |
| 1-Bromo-2-(methylsulfinyl)benzene | |
| CS(=O)C1=CC=CC=C1Br | |
| 219.096 | |
| 219.10 | |
| Liquid |
| Yellow | |
| C7H7BrOS | |
| 1 g | |
| NNNHMHLNXXGWBA-UHFFFAOYSA-N | |
| 1-bromo-2-methylsulfinylbenzene | |
| 11074873 | |
| ≥98.0% (GC) | |
| 2-Bromophenyl Methyl Sulfoxide |
Safety and Handling
TSCA : No