missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl Methyl Sulfoxide 98.0+%, TCI America™

Catalog No. B49905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49905G 5 g
B49901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49905G Supplier TCI America Supplier No. B49905G

Chemical Identifiers

CAS 7321-58-6
Molecular Formula C7H7BrOS
Molecular Weight (g/mol) 219.096
MDL Number MFCD18089235
InChI Key NNNHMHLNXXGWBA-UHFFFAOYSA-N
Synonym 1-Bromo-2-(methylsulfinyl)benzene
PubChem CID 11074873
IUPAC Name 1-bromo-2-methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 109°C
Quantity 5 g
Formula Weight 219.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromophenyl Methyl Sulfoxide