missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl isothiocyanate, 98%

Catalog No. p-7045251
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0997103 1 g
AAL0997106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0997103 Supplier Thermo Scientific Chemicals Supplier No. L0997103
Only null left

CAS: 13037-60-0 | C7H4BrNS | 214.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13037-60-0
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004799
InChI Key PAFORXDSYWMYGP-UHFFFAOYSA-N
PubChem CID 611654
IUPAC Name 1-bromo-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Br

Specifications

Density 1.591
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6843
Quantity 1 g
UN Number UN2922
Beilstein 2802536
Sensitivity Moisture sensitive
Synonym 2-bromophenyl isothiocyanate, benzene,1-bromo-2-isothiocyanato, benzene, 1-bromo-2-isothiocyanato, 2-bromophenylisothiocyanate, benzene, bromoisothiocyanato, isothiocyanic acid 2-bromophenyl ester, acmc-1bbbv, o-bromophenyl isothiocyanate, 1-bromo-2-isothiocyanato-benzene, #
Formula Weight 214.09
Percent Purity 98%
Chemical Name or Material 2-Bromophenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only