missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl Isocyanate 98.0+%, TCI America™

$124.89 - $473.48

Chemical Identifiers

CAS 1592-00-3
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00001995
InChI Key GOOVAYJIVMBWPP-UHFFFAOYSA-N
Synonym 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
PubChem CID 137097
IUPAC Name 1-bromo-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04985G
View Documents
TCI America
I04985G
5 g
Each for $124.89
Only null left
 
I049825G
View Documents
TCI America
I049825G
25 g
Each for $473.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1592-00-3
198.019
GOOVAYJIVMBWPP-UHFFFAOYSA-N
137097
C1=CC=C(C(=C1)N=C=O)Br
C7H4BrNO
MFCD00001995
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
1-bromo-2-isocyanatobenzene

Specifications

1592-00-3
C7H4BrNO
5 g
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
C1=CC=C(C(=C1)N=C=O)Br
198.019
198.02
Liquid
Yellow
MFCD00001995
2206
GOOVAYJIVMBWPP-UHFFFAOYSA-N
1-bromo-2-isocyanatobenzene
137097
≥98.0% (GC)
2-Bromophenyl Isocyanate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'