missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl isocyanate, 97%

Catalog No. AAL1252103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1252103 1 g
AAL1252106 5 g
AAL1252114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1252103 Supplier Thermo Scientific Chemicals Supplier No. L1252103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1592-00-3 | C7H4BrNO | 198.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1592-00-3
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00001995
InChI Key GOOVAYJIVMBWPP-UHFFFAOYSA-N
Synonym 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
PubChem CID 137097
IUPAC Name 1-bromo-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)Br

Specifications

Density 1.607
Boiling Point 91°C to 92°C (5 mmHg)
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.585
Quantity 1 g
UN Number UN2206
Beilstein 3588447
Sensitivity Moisture sensitive
Formula Weight 198.03
Percent Purity 97%
Chemical Name or Material 2-Bromophenyl isocyanate
Show More Show Less
Hazard Category H302+H312-H315-H319-H331-H334-H335
Hazard Statement GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only