missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenacyl Bromide 97.0+%, TCI America™
$262.83 - $915.25
Chemical Identifiers
| CAS | 49851-55-0 |
|---|---|
| Molecular Formula | C8H6Br2O |
| Molecular Weight (g/mol) | 277.943 |
| MDL Number | MFCD00044660 |
| InChI Key | LAXPJIJQTHJGCK-UHFFFAOYSA-N |
| PubChem CID | 2737522 |
| IUPAC Name | 2-bromo-1-(2-bromophenyl)ethanone |
| SMILES | C1=CC=C(C(=C1)C(=O)CBr)Br |
Chemical Identifiers
| 49851-55-0 | |
| 277.943 | |
| LAXPJIJQTHJGCK-UHFFFAOYSA-N | |
| 2-bromo-1-(2-bromophenyl)ethanone |
| C8H6Br2O | |
| MFCD00044660 | |
| 2737522 | |
| C1=CC=C(C(=C1)C(=O)CBr)Br |
Specifications
| 49851-55-0 | |
| 100°C | |
| MFCD00044660 | |
| 3265 | |
| LAXPJIJQTHJGCK-UHFFFAOYSA-N | |
| 2-bromo-1-(2-bromophenyl)ethanone | |
| 2737522 | |
| ≥97.0% (GC) | |
| 2-Bromophenacyl Bromide |
| Yellow | |
| C8H6Br2O | |
| 1 g | |
| 2,2′C-Dibromoacetophenone | |
| C1=CC=C(C(=C1)C(=O)CBr)Br | |
| 277.943 | |
| 277.94 | |
| Liquid |
Safety and Handling
TSCA : No