accessibility menu, dialog, popup

UserName

CAS RN 49851-55-0

IUPAC Name: 2-bromo-1-(2-bromophenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-bromophenyl)ethanone
Molecular Weight (g/mol): 277.94
Molecular Formula: C8H6Br2O
InChi Key: LAXPJIJQTHJGCK-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results