missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenacyl Bromide 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 100°C |
| Quantity | 5 g |
| UN Number | 3265 |
| Synonym | 2,2′-Dibromoacetophenone |
| Formula Weight | 277.94 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Bromophenacyl Bromide |
Safety and Handling
| TSCA | No |