missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromooctane (contains 3-Bromooctane) 85.0+%, TCI America™
Supplier: TCI America B134725G
Specifications
| 2-Bromooctane (contains 3-Bromooctane) | |
| Yellow | |
| C8H17Br | |
| 25 g | |
| FTJHYGJLHCGQHQ-UHFFFAOYSA-N | |
| 2-bromooctane | |
| 79046 | |
| ≥85.0% (GC) |
| 557-35-7 | |
| 191°C | |
| MFCD00040823 | |
| 2-Octyl Bromide | |
| CCCCCCC(C)Br | |
| 193.128 | |
| 193.13 | |
| Liquid |
Chemical Identifiers
| 557-35-7 | |
| 193.128 | |
| FTJHYGJLHCGQHQ-UHFFFAOYSA-N | |
| 79046 | |
| CCCCCCC(C)Br |
| C8H17Br | |
| MFCD00040823 | |
| 2-Octyl Bromide | |
| 2-bromooctane |
Safety and Handling
TSCA : Yes