missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)naphthalene, 96%

Catalog No. AC162561000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162561000 100 g Glass bottle
AC162560250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162561000 Supplier Thermo Scientific Chemicals Supplier No. 162561000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 939-26-4 | C11H9Br | 221.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939-26-4
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00004123
InChI Key RUHJZSZTSCSTCC-UHFFFAOYSA-N
Synonym 2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
PubChem CID 70320
IUPAC Name 2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 52°C to 57°C
Color Beige to Yellow
Boiling Point 213°C (100.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH2Br
Quantity 100 g
Beilstein 05,568
Formula Weight 221.1
Percent Purity 96%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)naphthalene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-359-5

RUO – Research Use Only