missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Bromomethyl)naphthalene, 96%
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (reacts), and chloroform.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (reacts), and chloroform.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 51°C to 55°C |
| Boiling Point | 213°C (100 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 636546 |
| Solubility Information | Soluble in water (reacts),and chloroform. |
| Formula Weight | 221.1 |
| Percent Purity | 96% |
| Chemical Name or Material | 2-(Bromomethyl)naphthalene |
Safety and Handling
| Hazard Category | H314-H317-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 213-359-5 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only