missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)acrylic Acid 98.0+%, TCI America™

Catalog No. B40481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40481G 1 g
B40485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40481G Supplier TCI America Supplier No. B40481G

Chemical Identifiers

CAS 72707-66-5
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.986
MDL Number MFCD00004259
InChI Key NOOYFQLPKUQDNE-UHFFFAOYSA-N
PubChem CID 558884
IUPAC Name 2-(bromomethyl)prop-2-enoic acid
SMILES C=C(CBr)C(=O)O

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 164.99
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)acrylic Acid
TSCA No
Recommended Storage Refrigerator