missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 90.0+%, TCI America™

Catalog No. p-7122802
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31991G 1 g
B31995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31991G Supplier TCI America Supplier No. B31991G

Chemical Identifiers

CAS 166330-03-6
Molecular Formula C7H14BBrO2
Molecular Weight (g/mol) 220.90
MDL Number MFCD09271779
InChI Key KBGMAXNDJAGTDD-UHFFFAOYSA-N
Synonym (Bromomethyl)boronic Acid Pinacol Ester
PubChem CID 10955110
IUPAC Name 2-(bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CBr)OC1(C)C

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
Formula Weight 220.90
Percent Purity ≥90.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator