accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CBr

CAS RN 166330-03-6

IUPAC Name: 2-(bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 220.9
Molecular Formula: C7H14BBrO2
InChi Key: KBGMAXNDJAGTDD-UHFFFAOYSA-N
SMILES: CC1(C)OB(CBr)OC1(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results