missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-1-(phenylsulfonyl)-1H-indole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC03308DA

 View more versions of this product

Catalog No. CC03308DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-(Bromomethyl)-1-(phenylsulfonyl)-1H-indole
96.5
97%
C15H12BrNO2S
1g
SYYPBRKVYRCLJS-UHFFFAOYSA-N
1-(benzenesulfonyl)-2-(bromomethyl)indole
2776225
97%
337508-54-0
100.0
Amber Glass Bottle
MFCD03086092
2-bromomethyl-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-2-bromomethyl indole, 1h-indole,2-bromomethyl-1-phenylsulfonyl
BrCC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
350.23
350.23

Chemical Identifiers

337508-54-0
350.23
SYYPBRKVYRCLJS-UHFFFAOYSA-N
2776225
BrCC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
C15H12BrNO2S
MFCD03086092
2-bromomethyl-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-2-bromomethyl indole, 1h-indole,2-bromomethyl-1-phenylsulfonyl
1-(benzenesulfonyl)-2-(bromomethyl)indole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions