missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,3-dioxolane, 97%

CAS: 4360-63-8 | C4H7BrO2 | 167 g/mol

$531.16 - $531.16

Chemical Identifiers

CAS 4360-63-8
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167
MDL Number MFCD00003214
InChI Key CKIIJIDEWWXQEA-UHFFFAOYSA-N
PubChem CID 78068
IUPAC Name 2-(bromomethyl)-1,3-dioxolane
SMILES C1COC(O1)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC209841000
SDS View Documents
Thermo Scientific Chemicals
209841000
100 mL Glass bottle
Each for $531.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4360-63-8
167
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-(bromomethyl)-1,3-dioxolane
C4H7BrO2
MFCD00003214
78068
C1COC(O1)CBr

Specifications

4360-63-8
100.0
1.6100g/mL
62°C
96% min. (GC)
C4H7BrO2
MFCD00003214
19, II, 8
2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
C1COC(O1)CBr
167
167
Liquid
96.0
Colorless
80°C to 82°C (27.0 mmHg)
Authentic
Glass bottle
1.4810 to 1.483
100 mL
1.61
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-(bromomethyl)-1,3-dioxolane
78068
97%
2-Bromomethyl-1, 3-dioxolane, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-443-6

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only