missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomalonaldehyde, 97+%

Catalog No. AC397900050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397900050 5 g Glass bottle
AC397900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397900050 Supplier Thermo Scientific Chemicals Supplier No. 397900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2065-75-0 | C3H3BrO2 | 150.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2065-75-0
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.95
InChI Key SURMYNZXHKLDFO-UHFFFAOYSA-N
Synonym 2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
PubChem CID 74945
IUPAC Name 2-bromopropanedial
SMILES C(=O)C(C=O)Br

Specifications

Melting Point 132.0°C to 136.0°C
Assay Percent Range 98.5 % min. (Acid)
Packaging Glass bottle
Linear Formula BrCH(CHO)2
Quantity 5 g
Formula Weight 150.95
Percent Purity 97+%
Chemical Name or Material 2-Bromomalonaldehyde
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only