missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisophthalaldehyde 98.0+%, TCI America™

Catalog No. B35685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35685G 5 g
B35681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35685G Supplier TCI America Supplier No. B35685G

Chemical Identifiers

CAS 79839-49-9
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD08461034
InChI Key RZUSSKMZLHKMHU-UHFFFAOYSA-N
Synonym 2-bromoisophthalaldehyde, 2-bromobenzene-1,3-dialdehyde, 2-bromo-1,3-diformylbenzene, 2,6-diformylbromobenzene, 2-bromo-benzene-1,3-dicarbaldehyde, 2-bromobenzene-1,3-dicarboxaldehyde, 1,3-diformyl-2-bromobenzene, 1,3-benzenedicarboxaldehyde, 2-bromo, acmc-209pia, ksc495c0d
PubChem CID 11435812
IUPAC Name 2-bromobenzene-1,3-dicarbaldehyde
SMILES BrC1=C(C=O)C=CC=C1C=O

Specifications

Melting Point 139°C
Quantity 5 g
Formula Weight 213.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoisophthalaldehyde