missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisophthalaldehyde, 97%

Catalog No. p-7055764
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6442303 1 g
AAH6442306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6442303 Supplier Thermo Scientific Chemicals Supplier No. H6442303

CAS: 79839-49-9 | C8H5BrO2 | 213.03 g/mol

Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 79839-49-9
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD08461034
InChI Key RZUSSKMZLHKMHU-UHFFFAOYSA-N
Synonym 2-bromoisophthalaldehyde, 2-bromobenzene-1,3-dialdehyde, 2-bromo-1,3-diformylbenzene, 2,6-diformylbromobenzene, 2-bromo-benzene-1,3-dicarbaldehyde, 2-bromobenzene-1,3-dicarboxaldehyde, 1,3-diformyl-2-bromobenzene, 1,3-benzenedicarboxaldehyde, 2-bromo, acmc-209pia, ksc495c0d
PubChem CID 11435812
IUPAC Name 2-bromobenzene-1,3-dicarbaldehyde
SMILES BrC1=C(C=O)C=CC=C1C=O

Specifications

Melting Point 139°C to 144°C
Quantity 1 g
Beilstein 4740331
Solubility Information Slightly soluble in water.
Formula Weight 213.03
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromoisophthalaldehyde
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only