missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisobutyric Acid 98.0+%, TCI America™

Catalog No. B060525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B060525G 25 g
B0605100G 100 g
B0605500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B060525G Supplier TCI America Supplier No. B060525G

Chemical Identifiers

CAS 2052-01-9
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004168
InChI Key XXSPGBOGLXKMDU-UHFFFAOYSA-N
Synonym 2-bromoisobutyric acid, 2-bromo-2-methylpropionic acid, isobromobutyric acid, propanoic acid, 2-bromo-2-methyl, alpha-bromoisobutyric acid, .alpha.-bromoisobutyric acid, 2-bromo-2-methyl-propanoic acid, propionic acid, 2-bromo-2-methyl, unii-821p2r6b0a, ccris 9046
PubChem CID 74924
IUPAC Name 2-bromo-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)Br

Specifications

Melting Point 46°C
Color White
Boiling Point 200°C
Quantity 25 g
UN Number 3261
Formula Weight 167.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoisobutyric Acid