missing translation for 'onlineSavingsMsg'
Learn More

2-Bromohexanoic Acid 98.0+%, TCI America™

Catalog No. B060125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B060125G 25 g
B0601500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B060125G Supplier TCI America Supplier No. B060125G

Chemical Identifiers

CAS 616-05-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004218
InChI Key HZTPKMIMXLTOSK-UHFFFAOYSA-N
Synonym 2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
PubChem CID 12013
IUPAC Name 2-bromohexanoic acid
SMILES CCCCC(C(=O)O)Br

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
UN Number 3265
Formula Weight 195.06
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromohexanoic Acid
EINECSNumber (2)-3249
TSCA Yes