accessibility menu, dialog, popup

UserName

H3CBrOHO

CAS RN 616-05-7

IUPAC Name: 2-bromohexanoic acid
Synonyms: α-bromo-n-caproic acid
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: HZTPKMIMXLTOSK-UHFFFAOYNA-N
SMILES: CCCCC(Br)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)